2,6-Diaminotoluene 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1,3-Diamino-2-methylbenzene, Toluene-2,6-diamine
164680250
164680050
ACRO164680250EA
145
EUR
ACRO164680250
ACRO164680050
2,6-Diaminotoluene 98%
2,6-Diaminotoluene
Formula:
C₇H₁₀N₂ MW: 122,17 g/mol Boiling Pt: 289 °C (760 mmHg) Melting Pt: 102…107 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007800 CAS Number: 823-40-5 EINECS: 212-513-9 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | Dark grey to brown to black Powder or pellets and/or chunks |
Infrared spectrum | Conforms |
Melting point | 102 to 107 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...