2,5-Dimethoxybenzyl alcohol 99%
Supplier: Thermo Scientific
182200250
182200050
ACRO182200250EA
155
EUR
ACRO182200250
ACRO182200050
2,5-Dimethoxybenzyl alcohol 99%
2,5-Dimethoxybenzyl alcohol
Formula:
C₉H₁₂O₃ MW: 168,19 g/mol Boiling Pt: 122…125 °C (1 mmHg) Flash Pt: >110 °C |
MDL Number:
MFCD00004615 CAS Number: 33524-31-1 EINECS: 251-562-0 |
Specification Test Results
Appearance | Clear colorless to light yellow Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5460 to 1.5480 (20°C, 589 nm) |
GC | ≥98.5 % |
Specific optical rotation | +68° to +78° (20°C, 589 nm) (1% in CHCl3) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...