3,4,5-Trimethoxybenzyl alcohol ≥98%

Supplier: Thermo Scientific

Synonyms: 2,4,6-Trimethoxybenzyl alcohol, (3,4,5-Trimethoxyphenyl)methanol

A13071.09 A13071.18 A13071.30
A13071.09EA 42.7 EUR
A13071.09 A13071.18 A13071.30
3,4,5-Trimethoxybenzyl alcohol ≥98%
3,4,5-Trimethoxybenzyl alcohol

Purity: 98% CAS No.: 3840-31-1 Molecular Formula: C10H14O4 Formula Weight: 198.22 Flash Point: 110°(230°F) Refractive Index: 1.5445 Boiling Point: 228°/25mm Beilstein Reference: 2104523 MDL No.: MFCD00004639

Formula: C₁₀H₁₄O₄
MW: 198,22 g/mol
Boiling Pt: 228 °C (25 mmHg)
Melting Pt: 36…38 °C
Density: 1,199
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060356
CAS Number: 3840-31-1
EINECS: 223-331-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR