4-Ethylresorcinol 98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Ethylbenzene-1,3-diol

118950050 118950250
ACRO118950050EA 45.2 EUR
ACRO118950050 ACRO118950250
4-Ethylresorcinol 98%
4-Ethylresorcinol
Formula: C₈H₁₀O₂
MW: 138,17 g/mol
Boiling Pt: 283 °C (1013 hPa)
Melting Pt: 95…99 °C
MDL Number: MFCD00002283
CAS Number: 2896-60-8
EINECS: 220-777-1

Order Now

Specification Test Results

Appearance Light beige Crystalline powder
Infrared spectrum Conforms
Melting point 95 to 99 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR