2-tert-Butylhydroquinone 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
t-Butylhydroquinone, 2-t-Butylhydroquinone, 2-(tert-Butyl)benzene-1,4-diol(may occur to produce black solid), tert-Butylhydroquinone, TBHQ
150820020
150820050
150822500
150825000
ACRO150820020EA
414
EUR
ACRO150820020
ACRO150820050
ACRO150822500
ACRO150825000
2-tert-Butylhydroquinone 97%
2-tert-Butylhydroquinone
Formula:
C₁₀H₁₄O₂ MW: 166.22 g/mol Boiling Pt: 273 Melting Pt: 125…130 °C Flash Pt: 171 Storage Temperature: Ambient |
MDL Number:
MFCD00002344 CAS Number: 1948-33-0 EINECS: 217-752-2 UN: 3077 ADR: 9 III |
Specification Test Results
Appearance | White to tan Crystalline powder (May contain black particles) |
Infrared spectrum | Conforms |
Melting point | 125 to 130 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...