3-Hydroxybenzyl alcohol ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (3-Hydroxyphenyl)methanol, KSD 2405, 3-(Hydroxymethyl)phenol

A15981.06 A15981.14 A15981.22
A15981.06EA 39.5 EUR
A15981.06 A15981.14 A15981.22
3-Hydroxybenzyl alcohol ≥98%
3-Hydroxybenzyl alcohol

Purity: 98% CAS No.: 620-24-6 Molecular Formula: C7H8O2 Formula Weight: 124.14 Melting Point: 69-72° Boiling Point: ca 300° dec. MDL No.: MFCD00004643

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: ∼300 °C (dec.)
Melting Pt: 69…72 °C
Density: 1.16
Storage Temperature: Ambient
MDL Number: MFCD00004643
CAS Number: 620-24-6
EINECS: 210-633-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR