Nonylbenzene ≥97%
Supplier: Thermo Fisher Scientific
A12957.09
A12957.18
A12957.30
A12957.09EA
47.2
EUR
A12957.09
A12957.18
A12957.30
Nonylbenzene ≥97%
Nonylbenzene
Purity: 97%
CAS No.: 1081-77-2
Molecular Formula: C15H24
Formula Weight: 204.36
Flash Point:
Formula:
C₁₅H₂₄ Boiling Pt: 281…283 °C Melting Pt: –24 °C Density: 0,858 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009575 CAS Number: 1081-77-2 EINECS: 214-103-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...