Bromocresol purple ACS, Reag. Ph. Eur., Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ200

1.03025.0005 1.03025.0025 1.03025.0500
1.03025.0005EA 97 EUR
1.03025.0005 1.03025.0025 1.03025.0500
Bromocresol purple ACS, Reag. Ph. Eur., Supelco®
Bromocresol purple
Formula: C₂₁H₁₆Br₂O₅S
Melting Pt: 242 °C
Storage Temperature: Ambient
MDL Number: MFCD00011681
CAS Number: 115-40-2
EINECS: 204-087-8
Merck Index: 13,01371

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 242 °C
Molar Mass 540.22 g/mol
Bulk Density 515 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR