Benzophenone 99+%, pure
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Diphenylmethanone, Diphenyl ketone
219680500
219680010
219680050
219682500
ACRO219680500EA
29.2
EUR
ACRO219680500
ACRO219680010
ACRO219680050
ACRO219682500
Benzophenone 99+%, pure
Benzophenone
Formula:
C₁₃H₁₀O MW: 182,22 g/mol Boiling Pt: 305,4 °C (1013 hPa) Melting Pt: 47…49 °C Density: 1,085 g/cm³ (20 °C) Flash Pt: 143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003076 CAS Number: 119-61-9 EINECS: 204-337-6 ADR: 9,III Merck Index: 13,01099 |
Specification Test Results
Appearance | White to off-white Crystalline powder or flakes |
Infrared spectrum | Conforms |
Melting point | 47 to 49 °C |
GC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...