2-Benzoylbenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A13281.0B
A13281.30
A13281.0BEA
0
EUR
A13281.0B
A13281.30
2-Benzoylbenzoic acid ≥98%
2-Benzoylbenzoic acid
Purity: 98+%
CAS No.: 85-52-9
Molecular Formula: C14H10O3
Formula Weight: 226.23
Odour: Odourless
Melting Point: 127-130°
Boiling Point: 257°
Beilstein Reference: 1107841
MDL No.: MFCD00002472
Formula:
C₁₄H₁₀O₃ Boiling Pt: 257 °C Melting Pt: 127…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002472 CAS Number: 85-52-9 EINECS: 201-612-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...