2-Hydroxybenzophenone 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(2-Hydroxyphenyl)(phenyl)methanone, o-Hydroxybenzophenone
146920100
146920250
146920010
ACRO146920100EA
108
EUR
ACRO146920100
ACRO146920250
ACRO146920010
2-Hydroxybenzophenone 99%
2-Hydroxybenzophenone
Formula:
C₁₃H₁₀O₂ MW: 198,22 g/mol Boiling Pt: 171…173 °C (12 mmHg) Melting Pt: 37…40 °C Density: 1,03 g/cm³ (172 °C) Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002216 CAS Number: 117-99-7 EINECS: 204-226-2 |
Specification Test Results
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 37 to 40 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...