2-Hydroxybenzophenone ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: (2-Hydroxyphenyl)(phenyl)methanone, o-Hydroxybenzophenone

L11100.06 L11100.14
L11100.06EA 49 EUR
L11100.06 L11100.14
2-Hydroxybenzophenone ≥99%
2-Hydroxybenzophenone

Purity: 99% CAS No.: 117-99-7 Molecular Formula: C13H10O2 Formula Weight: 198.22 Melting Point: 38-40° Boiling Point: 171-173°/12mm MDL No.: MFCD00002216

Formula: C₁₃H₁₀O₂
MW: 198,22 g/mol
Boiling Pt: 171…173 °C (12 mmHg)
Melting Pt: 38…40 °C
Density: 1,03 g/cm³ (172 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002216
CAS Number: 117-99-7
EINECS: 204-226-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR