3-Hydroxybenzophenone ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: m-Hydroxybenzophenone, (3-Hydroxyphenyl)(phenyl)methanone

A18061.03 A18061.06 A18061.14
A18061.03EA 66.5 EUR
A18061.03 A18061.06 A18061.14
3-Hydroxybenzophenone ≥99%
3-Hydroxybenzophenone

Purity: 99% CAS No.: 13020-57-0 Molecular Formula: C13H10O2 Formula Weight: 198.22 Melting Point: 113-115° MDL No.: MFCD00002297

Formula: C₁₃H₁₀O₂
MW: 198,22 g/mol
Boiling Pt: 148 °C (4 mmHg)
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00002297
CAS Number: 13020-57-0
EINECS: 235-879-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR