4-Benzyloxybenzyl alcohol ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: (4-(Benzyloxy)phenyl)methanol

A14949.09 A14949.18
A14949.09EA 45.2 EUR
A14949.09 A14949.18
4-Benzyloxybenzyl alcohol ≥98%
4-Benzyloxybenzyl alcohol

Purity: 98+% CAS No.: 836-43-1 Molecular Formula: C14H14O2 Formula Weight: 214.26 Melting Point: 84-88° Beilstein Reference: 1877819

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00004654
CAS Number: 836-43-1
EINECS: 212-649-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR