Trityl alcohol 97%
Supplier: Thermo Scientific
Synonyms:
Triphenylmethanol, Triphenylcarbinol
158910500
158912500
158910010
ACRO158910500EA
49.9
EUR
ACRO158910500
ACRO158912500
ACRO158910010
Trityl alcohol 97%
Trityl alcohol
Formula:
C₁₉H₁₆O MW: 260,34 g/mol Boiling Pt: 360 °C (760 mmHg) Melting Pt: 160…164 °C Density: 1,1884 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004445 CAS Number: 76-84-6 EINECS: 200-988-5 Merck Index: 13,09810 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 160 to 164 °C |
GC | ≥96.0 % |
Chloride (Cl) | ≤140 ppm |
Arsenic (As) | ≤8 ppm |
Heavy metals (as Pb) | ≤20 ppm |
Insoluble matter | ≤0.2 % (in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...