3,4-Dimethylphenylboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Dihydroxy(3,4-dimethylphenyl)borane, (3,4-Dimethylphenyl)metaboric acid, 3,4-Dimethylbenzeneboronic acid
L17461.03
L17461.06
L17461.03EA
40
EUR
L17461.03
L17461.06
3,4-Dimethylphenylboronic acid ≥98%
3,4-Dimethylphenylboronic acid
Purity: 98+%
CAS No.: 55499-43-9
Molecular Formula: C8H11BO2
Formula Weight: 149.99
Melting Point: ca 220°
Beilstein Reference: 2963858
Formula:
C₈H₁₁BO₂ MW: 149,98 g/mol Melting Pt: ∼ 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD01009694 CAS Number: 55499-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...