2,3-Dimethylbenzoyl chloride 96%
Supplier: Thermo Scientific
|
Danger
|
H25873.22
H25873.14
H25873.06
H25873.22EA
810
EUR
H25873.22
H25873.14
H25873.06
2,3-Dimethylbenzoyl chloride 96%
2,3-Dimethylbenzoyl chloride
Purity: 96%
CAS No.: 21900-46-9
Molecular Formula: C9H9ClO
Formula Weight: 168.62
Refractive Index: 1.5550
Melting Point: 2°
Boiling Point: 122-124°
UN No.: 3265
MDL No.: MFCD00045217
Formula:
C₉H₉ClO MW: 168,62 g/mol Boiling Pt: 122…124 °C Melting Pt: 2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045217 CAS Number: 21900-46-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...