2,3-Dimethylbenzamide ≥98%
Supplier: Thermo Scientific
|
Warning
|
A13585.03
A13585.06
A13585.03EA
22.3
EUR
A13585.03
A13585.06
2,3-Dimethylbenzamide ≥98%
2,3-Dimethylbenzamide
Purity: 98%
CAS No.: 5580-34-7
Molecular Formula: C9H11NO
Formula Weight: 149.19
Melting Point: 155-159°
Beilstein Reference: 2574226
Formula:
C₉H₁₁NO Melting Pt: 155…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017570 CAS Number: 5580-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...