2,5-Bis(trifluoromethyl)benzoic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

H26184.03 H26184.06
H26184.03EA 21.3 EUR
H26184.03 H26184.06
2,5-Bis(trifluoromethyl)benzoic acid ≥98%
2,5-Bis(trifluoromethyl)benzoic acid

Purity: 98% CAS No.: 42580-42-7 Molecular Formula: C9H4F6O2 Formula Weight: 258.12 Melting Point: 77-80° MDL No.: MFCD00013249

Formula: C₉H₄F₆O₂
MW: 258.12 g/mol
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00013249
CAS Number: 42580-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR