3,4-Dimethylphenyl isocyanate ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3,4-Xylyl isocyanate, 4-Isocyanato-1,2-dimethylbenzene, 3,4-Dimethylphenylisocyanate
L15998.03
L15998.06
L15998.14
L15998.03EA
29
EUR
L15998.03
L15998.06
L15998.14
3,4-Dimethylphenyl isocyanate ≥97%
3,4-Dimethylphenyl isocyanate
Purity: 97%
CAS No.: 51163-27-0
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 92°(197°F)
Refractive Index: 1.5380
Boiling Point: 85°/2mm
Beilstein Reference: 775149
UN No.: 2206
MDL No.: MFCD00013867
Formula:
C₉H₉NO MW: 147,18 g/mol Boiling Pt: 85 °C (2 mmHg) Density: 1,051 Flash Pt: 92 °C (197 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013867 CAS Number: 51163-27-0 UN: 2206 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...