4-Isopropylbenzyl alcohol 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
187800250
187801000
187805000
ACRO187800250EA
35.7
EUR
ACRO187800250
ACRO187801000
ACRO187805000
4-Isopropylbenzyl alcohol 97%
4-Isopropylbenzyl alcohol
Formula:
C₁₀H₁₄O Boiling Pt: 248 °C (760 mmHg) Melting Pt: 28 °C Density: 0,98 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004663 CAS Number: 536-60-7 EINECS: 208-640-4 Merck Index: 13,02646 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5180 to 1.5210 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...