4,4'-Di-tert-butylbiphenyl ≥99%
Supplier: Thermo Scientific
Synonyms:
1-tert-butyl-4-(4-tert-butylphenyl)benzene, 4,4'-Di-tert-butyl-1,1'-biphenyl
B21470.06
B21470.14
B21470.22
B21470.06EA
53.2
EUR
B21470.06
B21470.14
B21470.22
4,4'-Di-tert-butylbiphenyl ≥99%
4,4'-Di-tert-butylbiphenyl
Purity: 99%
CAS No.: 1625-91-8
Molecular Formula: C20H26
Formula Weight: 266.43
Melting Point: 126-129°
Boiling Point: 190-192°/13mm
MDL No.: MFCD00008834
Formula:
C₂₀H₂₆ MW: 266,43 g/mol Boiling Pt: 190…192 °C (13 mmHg) Melting Pt: 126…129 °C Density: 0,9691 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008834 CAS Number: 1625-91-8 EINECS: 216-615-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...