3-Bromobiphenyl (PBB No. 2) 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: PBB No. 2

339100010 339100050
ACRO339100010EA 67.3 EUR
ACRO339100010 ACRO339100050
3-Bromobiphenyl (PBB No. 2) 97%
3-Bromobiphenyl (PBB No. 2)
Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 299
Melting Pt: 9
Density: 1.398 g/cm³
Flash Pt: 110 °C
MDL Number: MFCD00000082
CAS Number: 2113-57-7
EINECS: 218-304-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6380 to 1.6420 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR