4-Fluorobiphenyl ≥97%
Supplier: Thermo Scientific
Synonyms:
PFB No. 3, 4-Fluoro-1,1-biphenyl
L07170.03
L07170.06
L07170.03EA
45.7
EUR
L07170.03
L07170.06
4-Fluorobiphenyl ≥97%
4-Fluorobiphenyl
Purity: 97+%
CAS No.: 324-74-3
Molecular Formula: C12H9F
Formula Weight: 172.20
Melting Point: 72-76°
Boiling Point: 253°
MDL No.: MFCD00011650
Formula:
C₁₂H₉F MW: 172,2 g/mol Boiling Pt: 253 °C Melting Pt: 72…76 °C Density: 1,247 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011650 CAS Number: 324-74-3 EINECS: 206-304-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...