2-Iodobiphenyl ≥98%

Supplier: Thermo Fisher Scientific

L06990.03 L06990.06
L06990.03EA 47.5 EUR
L06990.03 L06990.06
2-Iodobiphenyl ≥98%
2-Iodobiphenyl

Purity: 98% CAS No.: 2113-51-1 Molecular Formula: C12H9I Formula Weight: 280.11 Flash Point: 110°(230°F) Refractive Index: 1.6610 Boiling Point: 300° Beilstein Reference: 2206820 MDL No.: MFCD00039396

Formula: C₁₂H₉I
Boiling Pt: 300 °C
Density: 1,590
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039396
CAS Number: 2113-51-1
EINECS: 218-303-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR