p-Terphenyl 99+%, pure
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Phenyl-1,1'-biphenyl, 1,4-Diphenylbenzene, 4-Phenylbiphenyl, 1,4-Terphenyl
137920250
137921000
137925000
ACRO137920250EA
43.9
EUR
ACRO137920250
ACRO137921000
ACRO137925000
p-Terphenyl 99+%, pure
p-Terphenyl
Formula:
C₁₈H₁₄ MW: 230,31 g/mol Boiling Pt: 383 °C (760 mmHg) Melting Pt: 209…216 °C Density: 1,234 g/cm³ (0 °C) Flash Pt: 207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003061 CAS Number: 92-94-4 EINECS: 202-205-2 ADR: 9,III |
Specification Test Results
Appearance | White to light beige Glistening crystalline powder or crystals or flakes |
Infrared spectrum | Conforms |
Melting point | 211 to 216 °C |
GC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...