Coumarin for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1-Benzopyran-2-one, 2H-Chromen-2-one, 1,2-Benzopyrone
M-Clarity™ quality level = MQ200
8.22316.0250
8.22316.1000
8.22316.0250EA
64.7
EUR
8.22316.0250
8.22316.1000
Coumarin for synthesis, Sigma-Aldrich®
Coumarin
Formula:
C₉H₆O₂ MW: 146,15 g/mol Boiling Pt: 298 °C (1013 hPa) Melting Pt: 67…69 °C Density: 0,935 g/cm³ (20 °C) Flash Pt: 162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006850 CAS Number: 91-64-5 EINECS: 202-086-7 UN: 2811 ADR: 6.1,III Merck Index: 13,02588 |
Specification Test Results
Solubility | 1.7 g/l (20 °C) |
Melting Point | 68 - 71 °C |
Molar Mass | 146.14 g/mol |
Boiling Point | 298 - 302 °C |
Vapor Pressure | 1.3 hPa (106 °C) |
Flash Point | 162 °C |
Density | 0.935 g/cm3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...