Broxyquinoline ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5,7-Dibromo-8-hydroxyquinoline, 5,7-Dibromo-8-quinolinol

A14284.18 A14284.30
A14284.18EA 66.2 EUR
A14284.18 A14284.30
Broxyquinoline ≥98%
Broxyquinoline

Purity: 98% CAS No.: 521-74-4 Molecular Formula: C9H5Br2NO Formula Weight: 302.96 Melting Point: 198-203° Beilstein Reference: 153624 Merck Reference: 14,1453 MDL No.: MFCD00006785

Formula: C₉H₅Br₂NO
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 198…203 °C
Density: 2,189 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006785
CAS Number: 521-74-4
EINECS: 208-317-8
Merck Index: 14,1453

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR