Vitamin B12 (cyanocobalamin)

Supplier: Roth Carl

T915.1 T915.2
ROTHT915.1EA 67 EUR
ROTHT915.1 ROTHT915.2
Vitamin B12 (cyanocobalamin)
Vitamin B12 (cyanocobalamin)
Formula: C₆₃H₈₈CoN₁₄O₁₄P
Melting Pt: 320 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151092
CAS Number: 68-19-9
EINECS: 200-680-0
Merck Index: 12,10152

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR