p-Benzoquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
p-Quinone, Quinone, 1,4-Cyclohexadiene-3,6-dione, 1,4-Benzoquinone
M-Clarity™ quality level = MQ200
8.02410.0100
8.02410.1000
8.02410.9050
8.02410.9025
8.02410.0100EA
30
EUR
8.02410.0100
8.02410.1000
8.02410.9050
8.02410.9025
p-Benzoquinone for synthesis, Sigma-Aldrich®
p-Benzoquinone
Formula:
C₆H₄O₂ Boiling Pt: ∼174 °C (1013 hPa) Melting Pt: 112…114 °C Density: 1,318 g/cm³ (20 °C) Flash Pt: 77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001591 CAS Number: 106-51-4 EINECS: 203-405-2 UN: 2587 ADR: 6.1,II Merck Index: 12,08259 |
|
Specification Test Results
Solubility | 10 g/l (25 °C) |
Melting Point | 110 - 113 °C |
Molar Mass | 108.09 g/mol |
Bulk Density | 700 kg/m3 |
Boiling Point | 180 °C (sublimed) |
Vapor Pressure | 0.12 hPa (20 °C) |
Density | 1.32 g/cm3 (20 °C) |
pH | 4 (1 g/l, H2O, 20 °C) |
Ignition Point | 560 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...