Cyclopentanecarbonyl chloride 98%

Supplier: Thermo Fisher Scientific
Danger

354370050 354370250 354371000
ACRO354370050EA 75.2 EUR
ACRO354370050 ACRO354370250 ACRO354371000
Cyclopentanecarbonyl chloride 98%
Cyclopentanecarbonyl chloride
Formula: C₆H₉ClO
Boiling Pt: 54…56 °C (15 mmHg)
Density: 1,091 g/cm³
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00001370
CAS Number: 4524-93-0
EINECS: 224-856-1
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4640 (20°C, 589 nm)
Titration RCOCl ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR