Cyclohexanol

Supplier: Roth Carl
Warning

7756.1 7756.2
ROTH7756.1EA 0 EUR
ROTH7756.1 ROTH7756.2
Cyclohexanol
Cyclohexanol
Formula: C₆H₁₂O
Boiling Pt: 161 °C (1013 hPa)
Melting Pt: 25 °C
Density: 0,947 g/cm³ (20 °C)
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00003855
CAS Number: 108-93-0
EINECS: 203-630-6
Merck Index: 13,02754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8878907005758628f5927d7b0f43a99f57de5ae8&vid=c5d64c7b5d58d205d87b42eb97190b1693d3508a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR