Ethynylcyclohexane 98%

Supplier: Thermo Scientific
Danger

380790010 380790050
ACRO380790010EA 59.4 EUR
ACRO380790010 ACRO380790050
Ethynylcyclohexane 98%
Ethynylcyclohexane
Formula: C₈H₁₂
Boiling Pt: 130…132 °C (760 mmHg)
Density: 0,828 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00001513
CAS Number: 931-48-6
EINECS: 213-236-6
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4530 to 1.4550 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR