2-Cyclohexylethanol ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
B20782.0B
B20782.18
B20782.30
B20782.0BEA
498
EUR
B20782.0B
B20782.18
B20782.30
2-Cyclohexylethanol ≥99%
2-Cyclohexylethanol
Purity: 99%
CAS No.: 4442-79-9
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 86°(186°F)
Refractive Index: 1.4660
Melting Point: -20°
Boiling Point: 206-207°
Beilstein Reference: 1848152
MDL No.: MFCD00001525
Formula:
C₈H₁₆O Boiling Pt: 206…207 °C Melting Pt: –20 °C Density: 0,920 Flash Pt: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001525 CAS Number: 4442-79-9 EINECS: 224-672-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...