Cyclobutylméthanol ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Cyclobutanemethanol, Cyclobutylmethanol, Hydroxymethylcyclobutane

B21480.06 B21480.14
B21480.06EA 38.4 EUR
B21480.06 B21480.14
Cyclobutylméthanol ≥99%
Cyclobutylméthanol

Purity: 99% CAS No.: 4415-82-1 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 39°(102°F) Refractive Index: 1.4465 Boiling Point: 143-144° Beilstein Reference: 2036027 UN No.: 1105 MDL No.: MFCD00001330

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 143…144 °C
Density: 0.913
Flash Pt: 39 °C (102 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001330
CAS Number: 4415-82-1
EINECS: 224-575-4
UN: 1105
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR