1,3,5,7-Cyclooctatetraene ≥98% stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Cyclooctatetraene, COT
Stabilisation:
Stabilised with IRGANOX 1076
B22636.06
B22636.14
B22636.06EA
143
EUR
B22636.06
B22636.14
1,3,5,7-Cyclooctatetraene ≥98% stabilised
1,3,5,7-Cyclooctatetraene
Stabilized with 0.1% Hydroquinone
Purity: 98%
CAS No.: 629-20-9
Molecular Formula: C8H8
Formula Weight: 104.15
Storage Temperature: 2°C to 8°C
Flash Point: 20°(68°F)
Refractive Index: 1.537
Odour: Unpleasant
Melting Point: -5°
Boiling Point: 142-143°
Beilstein Reference: 2496800
UN No.: 2358
MDL No.: MFCD00004161
Formula:
C₈H₈ Boiling Pt: 142…143 °C Melting Pt: –5 °C Density: 0,925 Flash Pt: 20 °C (68 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004161 CAS Number: 629-20-9 EINECS: 211-080-3 UN: 2358 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...