α-Tropolone 98%
Supplier: Thermo Fisher Scientific
Synonyms:
Tropolone, 2-Hydroxytropone, 2-Hydroxycyclohepta-2,4,6-trien-1-one, 1,2-Tropolone, 2-Hydroxycyclohepta-2,4,6-trienone, α-Tropolone
140580010
140580050
140580250
ACRO140580010EA
51.9
EUR
ACRO140580010
ACRO140580050
ACRO140580250
α-Tropolone 98%
α-Tropolone
Formula:
C₇H₆O₂ MW: 122.12 g/mol Boiling Pt: 115 °C (12 mmHg) Melting Pt: 50…54 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004158 CAS Number: 533-75-5 EINECS: 208-577-2 |
Specification Test Results
Appearance | Off-white to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 50 to 54 °C (on dry substance) |
GC | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...