3-Methyl-2-cyclohexen-1-one ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Methyl-2-cyclohexenone, 3-Methylcyclohex-2-enone
A15704.09
A15704.14
A15704.18
A15704.22
A15704.09EA
84.3
EUR
A15704.09
A15704.14
A15704.18
A15704.22
3-Methyl-2-cyclohexen-1-one ≥98%
3-Methyl-2-cyclohexen-1-one
Purity: 98%
CAS No.: 1193-18-6
Molecular Formula: C7H10O
Formula Weight: 110.15
Flash Point: 68°(154°F)
Refractive Index: 1.4940
Odour: Pleasant
Boiling Point: 199-200°
Beilstein Reference: 1560601
MDL No.: MFCD00001581
Formula:
C₇H₁₀O MW: 110,16 g/mol Boiling Pt: 199…200 °C Melting Pt: 198…199 °C Density: 0,971 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001581 CAS Number: 1193-18-6 EINECS: 214-769-7 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...