Barbituric acid
Supplier: LGC Standards PROMOCHEM
Synonyms:
N,N'-Malonylurea
, 2,4,6-Trihydroxypyrimidine
PROOMM0265.03EA
559
EUR
PROOMM0265.03
Barbituric acid
Barbituric acid
Formula:
C₄H₄N₂O₃ MW: 128,09 g/mol Boiling Pt: 441 °C (1013 hPa) Melting Pt: 250…252 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006666 CAS Number: 67-52-7 EINECS: 200-658-0 Merck Index: 13,00966 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...