9,10-Anthraquinone ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Anthracene-9,10-dione, Anthraquinone

A12735.30
A12735.30EA 71 EUR
A12735.30
9,10-Anthraquinone ≥98%
9,10-Anthraquinone

Purity: 98+% CAS No.: 84-65-1 Molecular Formula: C14H8O2 Formula Weight: 208.22 Flash Point: 185°(365°F) Odour: Odourless Boiling Point: 377° Beilstein Reference: 390030 Merck Reference: 14,687 MDL No.: MFCD00001188

Formula: C₁₄H₈O₂
MW: 208,22 g/mol
Boiling Pt: 377 °C
Melting Pt: 283…287 °C
Density: 1,438
Flash Pt: 185 °C (365 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001188
CAS Number: 84-65-1
EINECS: 201-549-0
Merck Index: 14,687

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR