Fluorene ≥98%
Supplier: Thermo Scientific
Synonyms:
9H-Fluorene, Diphenylenmethane
L02124.14
L02124.22
L02124.36
L02124.14EA
28.3
EUR
L02124.14
L02124.22
L02124.36
Fluorene ≥98%
Fluorene
Purity: 98+%
CAS No.: 86-73-7
Molecular Formula: C13H10
Formula Weight: 166.22
Flash Point: 151°(303°F)
Melting Point: 113-117°
Boiling Point: 295°
Beilstein Reference: 1363491
Merck Reference: 14,4155
UN No.: 3077
MDL No.: MFCD00001111
Formula:
C₁₃H₁₀ MW: 166,22 g/mol Boiling Pt: 295 °C Melting Pt: 113…117 °C Density: 1,202 Flash Pt: 151 °C (303 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001111 CAS Number: 86-73-7 EINECS: 201-695-5 UN: 3077 ADR: 9,III Merck Index: 14,4155 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...