2-Ethyl-9,10-anthraquinone ≥98%

Supplier: Thermo Scientific
Warning

L07206.36 L07206.06
L07206.36EA 0 EUR
L07206.36 L07206.06
2-Ethyl-9,10-anthraquinone ≥98%
2-Ethyl-9,10-anthraquinone

Purity: 98% CAS No.: 84-51-5 Molecular Formula: C16H12O2 Formula Weight: 236.27 Flash Point: 210°(410°F) Melting Point: 107-111° Boiling Point: 180-190° Beilstein Reference: 1969873 MDL No.: MFCD00001237

Formula: C₁₆H₁₂O₂
Boiling Pt: 180…190 °C
Melting Pt: 107…111 °C
Flash Pt: >210 °C (410 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001237
CAS Number: 84-51-5
EINECS: 201-535-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR