Xanthene-9-carboxylic acid ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
L10384.14
L10384.22
L10384.06
L10384.14EA
58
EUR
L10384.14
L10384.22
L10384.06
Xanthene-9-carboxylic acid ≥98%
Xanthene-9-carboxylic acid
Purity: 98%
CAS No.: 82-07-5
Molecular Formula: C14H10O3
Formula Weight: 226.23
Melting Point: ca 217° dec.
Beilstein Reference: 174414
Formula:
C₁₄H₁₀O₃ Melting Pt: ∼ 217 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005059 CAS Number: 82-07-5 EINECS: 201-394-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...