Fluorescein disodium salt

Supplier: Roth Carl

5283.1 5283.2
ROTH5283.1EA 0 EUR
ROTH5283.1 ROTH5283.2
Fluorescein disodium salt
Fluorescein disodium salt
Formula: C₂₀H₁₀Na₂O₅
Melting Pt: > 300 °C
Density: 1,601 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167039
CAS Number: 518-47-8
EINECS: 208-253-0
Merck Index: 13,04185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR