5-Methyl-2-furaldehyde ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A13264.22
A13264.14
A13264.22EA
116
EUR
A13264.22
A13264.14
5-Methyl-2-furaldehyde ≥98%
5-Methyl-2-furaldehyde
Purity: 98%
CAS No.: 620-02-0
Molecular Formula: C6H6O2
Formula Weight: 110.11
Flash Point: 72°(161°F)
Refractive Index: 1.5310
Boiling Point: 186-188°
Beilstein Reference: 106895
MDL No.: MFCD00003232
Formula:
C₆H₆O₂ Boiling Pt: 186…188 °C Density: 1,106 Flash Pt: 72 °C (161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003232 CAS Number: 620-02-0 EINECS: 210-622-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...