1,1,3,3-Tetramethylguanidine ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
A12314.AC
A12314.AE
A12314.AP
A12314.ACEA
28.9
EUR
A12314.AC
A12314.AE
A12314.AP
1,1,3,3-Tetramethylguanidine ≥99%
1,1,3,3-Tetramethylguanidine
Purity: 99%
CAS No.: 80-70-6
Molecular Formula: C5H13N3
Formula Weight: 115.18
Flash Point: 60°(140°F)
Refractive Index: 1.4690
Melting Point:
Formula:
C₅H₁₃N₃ Boiling Pt: 162 °C Melting Pt: < –30 °C Density: 0,918 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008323 CAS Number: 80-70-6 EINECS: 201-302-7 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...