Methylphosphonic acid 98%
Supplier: Thermo Fisher Scientific
|
Danger
|
354330050
354330250
354331000
ACRO354330050EA
78.5
EUR
ACRO354330050
ACRO354330250
ACRO354331000
Methylphosphonic acid 98%
Methylphosphonic acid
Formula:
CH₅O₃P Melting Pt: 103…107 °C Density: 1,639 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002137 CAS Number: 993-13-5 EINECS: 213-607-2 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 103 to 107 °C |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...