Sodium chlorite

Supplier: Roth Carl
Danger

4352.1 4352.2 4352.3
ROTH4352.1EA 0 EUR
ROTH4352.1 ROTH4352.2 ROTH4352.3
Sodium chlorite
Sodium chlorite
Formula: ClNaO₂
Boiling Pt: 112 °C (1013 hPa)
Melting Pt: 250…257 °C
Density: 1,28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003478
CAS Number: 7758-19-2
EINECS: 231-836-6
UN: 1496
ADR: 5.1,II
Merck Index: 13,08672

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR