Sodium sulphite

Supplier: Roth Carl

8637.1 8637.2
ROTH8637.1EA 0 EUR
ROTH8637.1 ROTH8637.2
Sodium sulphite
Sodium sulphite
Formula: Na₂SO₃
Melting Pt: > 500 °C
Density: 2,63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR