Potassium hydrogen sulphate
Supplier: Roth Carl
|
Danger
|
Synonyms:
Potassium bisulphate
P020.1
P020.2
ROTHP020.1EA
46.4
EUR
ROTHP020.1
ROTHP020.2
Potassium hydrogen sulphate
Potassium hydrogen sulphate
Formula:
KHSO₄ MW: 136.17 g/mol Melting Pt: 214 Density: 2.32 g/cm³ (20 °C) |
MDL Number:
MFCD00011404 CAS Number: 7646-93-7 EINECS: 231-594-1 UN: 2509 ADR: 8 II Merck Index: 13,07695 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...